Learn More
Bioworld Murashige and Skoog(MS) Modified Medium w/FeNaEDTA, 25 L
Catalog No. 50254996
Encompass_Preferred
Murashige & Skoog (MS) Modified Medium with FeNaEDTAContains the Macronutrients, Micronutrients as described by Murashige & Skoog (1962), FeNaEDTA in place of FeSO4 and NaEDTACytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 4.3 g/LpH: 4.4−5.2Soluble in water
Catalog No. 50-254-996 Supplier Bioworld Supplier No. 306301992
Murashige & Skoog (MS) Modified Medium with FeNaEDTAContains the Macronutrients, Micronutrients as described by Murashige & Skoog (1962), FeNaEDTA in place of FeSO4 and NaEDTACytokines, Sucrose, and Auxins can be added as needed Sold in individual containers with enough medium to make 10L, 25L, or 50L of mediumPlant tissue culture testedWorking Concentration: 4.3 g/LpH: 4.4−5.2Soluble in water