Learn More
Bioworld Murashige and Skoog(MS) Shoot Multiplication B Medium, 25 L
Catalog No. 50254723
Encompass_Preferred
Murashige & Skoog (MS) Shoot Multiplication B MediumContains Macronutrients, Micronutrients, and Vitamins as described by Murashige & Skoog (1967)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-723 Supplier Bioworld Supplier No. 306300793
Murashige & Skoog (MS) Shoot Multiplication B MediumContains Macronutrients, Micronutrients, and Vitamins as described by Murashige & Skoog (1967)Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested