Learn More
Bioworld Murashige and Skoog(MS) Stock Solution (10X), Micronutrient Only, 1 L
Catalog No. 50254821
Encompass_Preferred
Murashige & Skoog (MS) Stock Solution (10X)Contains only the Micronutrients as described by Murashige and Skoog (1962) in 10X SolutionPlant tissue culture testedWorking Concentration: 100 mL/LpH: 2.5–3.5Miscible in water
Catalog No. 50-254-821 Supplier Bioworld Supplier No. 306301282
Murashige & Skoog (MS) Stock Solution (10X)Contains only the Micronutrients as described by Murashige and Skoog (1962) in 10X SolutionPlant tissue culture testedWorking Concentration: 100 mL/LpH: 2.5–3.5Miscible in water