Learn More
Bioworld Murashige and Skoog(MS) Stock Solution (10X), Macronutrient Only, 1 L
Catalog No. 50254823
Encompass_Preferred
Murashige & Skoog (MS) Macronutrient Stock Solution (10X)Containsonly the Macronutrients as described by Murashige and Skoog (1962) in 10X SolutionPlant tissue culture testedWorking Concentration: 100 mL/LpH: 3.75–4.75Miscible with Water
Catalog No. 50-254-823 Supplier Bioworld Supplier No. 306301292
Murashige & Skoog (MS) Macronutrient Stock Solution (10X)Containsonly the Macronutrients as described by Murashige and Skoog (1962) in 10X SolutionPlant tissue culture testedWorking Concentration: 100 mL/LpH: 3.75–4.75Miscible with Water