Learn More
Bioworld Murashige and Skoog(MS) Vitamin Powder Mix, 100 mL
Catalog No. 50254994
Encompass_Preferred
Murashige & Skoog (MS) Vitamin Powder MixContains the Vitamins as described by Murashige & Skoog (1962)Plant tissue culture testedContains: 10.31g to make 100ml of 1000x stock solution. Working Concentration: Use 1ml of 1000x stock per liter of water or medium.pH: 3.5–4.5Soluble in waterContains: Glycine: 2mg/LMyo-Inositol: 100.00 mg/LNicotinic Acid: 0.50 mg/LPyridoxine HCl: 0.50 mg/LThiamine HCl: 0.10 mg/L
Catalog No. 50-254-994 Supplier Bioworld Supplier No. 306301981
Murashige & Skoog (MS) Vitamin Powder MixContains the Vitamins as described by Murashige & Skoog (1962)Plant tissue culture testedContains: 10.31g to make 100ml of 1000x stock solution. Working Concentration: Use 1ml of 1000x stock per liter of water or medium.pH: 3.5–4.5Soluble in waterContains: Glycine: 2mg/LMyo-Inositol: 100.00 mg/LNicotinic Acid: 0.50 mg/LPyridoxine HCl: 0.50 mg/LThiamine HCl: 0.10 mg/L