Learn More
Research Products International Corp Murashige & Skoog Modified Medium, NH4NO3 Free, 13.5 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 50213437
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962). Modified by eliminating ammonium nitrate.
  • Concentrate: 2652.09 mg/L
  • Hygroscopic
Catalog No. 50-213-437 Supplier Research Products International Corp Supplier No. M106005.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962). Modified by eliminating ammonium nitrate.
  • Concentrate: 2652.09 mg/L
  • Hygroscopic