Learn More
Research Products International Corp Murashige & Skoog Modified Medium, NH4NO3 Free, 27 Grams of Powder, Makes 10 Liters of Solution
SDP
Catalog No. 50213435
Encompass_Preferred
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962). Modified by eliminating ammonium nitrate.
  • Concentrate: 2652.09 mg/L
  • Hygroscopic
Catalog No. 50-213-435 Supplier Research Products International Corp Supplier No. M1060010.0
Basal salt mixture containing micro and macro elements with vitamins as described by Murashige and Skoog (1962). Modified by eliminating ammonium nitrate.
  • Concentrate: 2652.09 mg/L
  • Hygroscopic