Learn More
Research Products International Corp Murashige & Skoog Vitamin Mixture, 10.31 Grams of Powder, Makes 100 Milliliters of Solution
SDP
Catalog No. 50213490
Encompass_Preferred
Powdered vitamin mixture as described by Murashige & Skoog (1962). Makes 1000X stock solution. Package contains 103.1 grams of powder per liter of medium. Use at a concentration of 1 ml per liter to achieve final concentration. Hygroscopic.
Catalog No. 50-213-490 Supplier Research Products International Corp Supplier No. M701000.1
Powdered vitamin mixture as described by Murashige & Skoog (1962). Makes 1000X stock solution. Package contains 103.1 grams of powder per liter of medium. Use at a concentration of 1 ml per liter to achieve final concentration. Hygroscopic.