Learn More
Research Products International Corp Murashige & Skoog Vitamin Mixture, 25.80 Grams of Powder, Makes 250 Milliliters of Solution
SDP
Supplier:  Research Products International Corp M701000.25
Encompass_Preferred
Powdered vitamin mixture as described by Murashige & Skoog (1962). Makes 1000X stock solution. Package contains 103.1 grams of powder per liter of medium. Use at a concentration of 1 ml per liter to achieve final concentration. Hygroscopic.
Catalog No. 50-213-491