Learn More
Alkali Scientific Murashige & Skoog Vitamin Mixture, 10.31 Grams of Powder, Makes 100 Milliliters of Solution
SDP
Catalog No. 5000659747
Encompass_Preferred
Murashige & Skoog Vitamin Mixture, 10.31 Grams of Powder, Makes 100 Milliliters of Solution, is a key component in plant tissue culture applications. This vitamin mixture is a part of the Murashige and Skoog (MS) medium and provides essential vitamins that promote plant cell division, growth, and differentiation in vitro. It contains a mix of vital plant growth vitamins, which are crucial for various tissue culture applications, including somatic embryogenesis, organogenesis, and callus culture. This vitamin mixture is typically used alongside other MS medium components to create a complete growth environment for plant cells in laboratory experiments.
Catalog No. 50-006-59747 Supplier Alkali Scientific Supplier No. GES001CP
Murashige & Skoog Vitamin Mixture, 10.31 Grams of Powder, Makes 100 Milliliters of Solution, is a key component in plant tissue culture applications. This vitamin mixture is a part of the Murashige and Skoog (MS) medium and provides essential vitamins that promote plant cell division, growth, and differentiation in vitro. It contains a mix of vital plant growth vitamins, which are crucial for various tissue culture applications, including somatic embryogenesis, organogenesis, and callus culture. This vitamin mixture is typically used alongside other MS medium components to create a complete growth environment for plant cells in laboratory experiments.