Learn More
Research Products International Corp Murashige & Skoog Medium with MES Buffer, 24 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 50213498
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Murashige and Skoog (1962). MES buffer 500 mg/L added to prevent acidification during cultivation. Supplied as powder.
  • Concentration: 4802.1 mg/L
  • Hygroscopic
Catalog No. 50-213-498 Supplier Research Products International Corp Supplier No. M703005.0
Basal salt mixture containing micro and macro elements as described by Murashige and Skoog (1962). MES buffer 500 mg/L added to prevent acidification during cultivation. Supplied as powder.
  • Concentration: 4802.1 mg/L
  • Hygroscopic