Learn More
Research Products International Corp Murashige & Skoog Medium with Nitschs' Vitamins, 44 Grams of Powder, Makes 10 Liters of Solution
SDP
Catalog No. 50213493
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described Murashige and Skoog (1962) and vitamins as described by Nitsch & Nitsch (1969). Supplied as powder.
  • Concentration: 4410.64 mg/L
  • Hygroscopic
Catalog No. 50-213-493 Supplier Research Products International Corp Supplier No. M7020010.0
Basal salt mixture containing micro and macro elements as described Murashige and Skoog (1962) and vitamins as described by Nitsch & Nitsch (1969). Supplied as powder.
  • Concentration: 4410.64 mg/L
  • Hygroscopic