Learn More
Research Products International Corp Murashige & Skoog Medium with Nitschs' Vitamins, 4.4 Grams of Powder, Makes 1 Liter of Solution
SDP
Catalog No. 50213492
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described Murashige and Skoog (1962) and vitamins as described by Nitsch & Nitsch (1969). Supplied as powder.
  • Concentration: 4410.64 mg/L
  • Hygroscopic
Catalog No. 50-213-492 Supplier Research Products International Corp Supplier No. M702001.0
Basal salt mixture containing micro and macro elements as described Murashige and Skoog (1962) and vitamins as described by Nitsch & Nitsch (1969). Supplied as powder.
  • Concentration: 4410.64 mg/L
  • Hygroscopic