Learn More
Research Products International Corp Murashige & Skoog Medium with Nitschs' Vitamins, 22 Grams of Powder, Makes 5 Liters of Solution
SDP
Catalog No. 50213494
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described Murashige and Skoog (1962) and vitamins as described by Nitsch & Nitsch (1969). Supplied as powder.
Concentration: 4410.64 mg/L
Hygroscopic
Catalog No. 50-213-494 Supplier Research Products International Corp Supplier No. M702005.0
Basal salt mixture containing micro and macro elements as described Murashige and Skoog (1962) and vitamins as described by Nitsch & Nitsch (1969). Supplied as powder.
Concentration: 4410.64 mg/L
Hygroscopic