Learn More
Research Products International Corp Murashige and Skoog Medium with Vitamins, Van der Salm Modification, 44 Grams of Powder, Makes 10 Liters of Solution
SDP
Catalog No. 50213501
Encompass_Preferred
Basal salt mixture containing micro and macro elements as described by Murashige & Skoog (1962). Modified by replacing FeNaEDTA with FeEDDHA as described by Van der Salm et. al. (1994).
  • Concentration: 4464.49 mg/L
  • Hygroscopic
Catalog No. 50-213-501 Supplier Research Products International Corp Supplier No. M7040010.0
Basal salt mixture containing micro and macro elements as described by Murashige & Skoog (1962). Modified by replacing FeNaEDTA with FeEDDHA as described by Van der Salm et. al. (1994).
  • Concentration: 4464.49 mg/L
  • Hygroscopic