Learn More
Cayman Chemical Muraya pnculta Murayon 10mg
SDP
Catalog No. 502912252
Encompass_Preferred
A coumarin with antiplatelet activity; inhibits ADP-induced platelet aggregation in vitro
Catalog No. 50-291-2252 Supplier Cayman Chemical Supplier No. 3476710 MG
A coumarin with antiplatelet activity; inhibits ADP-induced platelet aggregation in vitro