Learn More
Medchemexpress LLC Murexide (ammonium purpurate) | 3051-09-0 | MFCD00012777 | 98.0% | 284.19 g/mol | C8H8N6O6 | 25G
SDP
Catalog No. 5000238314
Encompass_Preferred
Murexide (ammonium purpurate) is an analytical reagent used primarily as an indicator in complexometric titrations for determining water hardness and detecting certain metal ions. It is supplied as a solid powder with a typical purity of 98.0% and has the chemical formula C8H8N6O6 (MW 284.19 g/mol), making it suitable for laboratory and research analytical applications.
  • Indicator for complexometric titrations to determine water hardness.
  • Solid powder form suitable for analytical preparations.
  • Typical purity 98.0%.
  • Chemical formula C8H8N6O6; molecular weight 284.19 g/mol.
  • Available in multiple gram pack sizes for laboratory use.
Catalog No. 50-002-38314 Supplier Medchemexpress LLC Supplier No. HY12592925G
Murexide (ammonium purpurate) is an analytical reagent used primarily as an indicator in complexometric titrations for determining water hardness and detecting certain metal ions. It is supplied as a solid powder with a typical purity of 98.0% and has the chemical formula C8H8N6O6 (MW 284.19 g/mol), making it suitable for laboratory and research analytical applications.
  • Indicator for complexometric titrations to determine water hardness.
  • Solid powder form suitable for analytical preparations.
  • Typical purity 98.0%.
  • Chemical formula C8H8N6O6; molecular weight 284.19 g/mol.
  • Available in multiple gram pack sizes for laboratory use.