Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murexide Indicator, 0.2% (w/w) in Sodium Chloride, Powder Mixture, For Calcium, APHA, Spectrum™ Chemical
SDP

Catalog No. 18611833 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-611-833 500 g
18-611-832 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-611-833 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M280500GM
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Murexide Indicator
CAS 3051-09-0, 7647-14-5
Concentration 0.2 Percent (w/w)
Packaging Poly Bottle
Color Purple/Red to Red
Quantity 500 g
Physical Form Chemical Solution
Grade APHA

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.