Learn More
Murexide-Sodium Chloride Water Hardness Indicator, Reagents
Specifications
Specifications
| Chemical Name or Material | Murexide-Sodium Chloride Water Hardness Indicator |
| CAS | 7647-14-5, 3051-09-0 |
| Boiling Point | 1461°C |
| Packaging | Glass Bottle |
| Specific Gravity | 2.17 |
| Synonym | Ammonium Purpurate, Color Index 56085 |
| pH | 5 to 9 |
| Quantity | 100 g |
| Physical Form | Solid |
| Grade | ACS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.