Learn More
Medchemexpress LLC ω-Muricholic acid | 6830-03-1 | MFCD11142482 | ≥99.0% | 408.57 g/mol | C24H40O5 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1251381MG
Encompass_Preferred
ω-Muricholic acid (ω-MCA) is a murine-specific secondary bile acid supplied as a high-purity analytical reagent for biochemical and metabolic research. It is used in studies of bile acid metabolism, microbiome-host interactions, and metabolite profiling. Molecular formula C24H40O5, molecular weight 408.57 g/mol. Supplied for in vitro and analytical workflows; for research use only.
  • High-purity analytical standard for metabolite profiling.
  • Relevant to studies of bile acid metabolism and metabolic disease.
  • Molecular formula C24H40O5 and molecular weight 408.57 g/mol.
  • Offered in small, accurately measured quantities for analytical use.
  • For research use only; not for human or veterinary use.
Catalog No. 50-002-38187