Learn More
Cayman Chemical Murideoxycholic Acid, Purity: ≥95%, CAS: 668-49-5, 1 mg
SDP
Catalog No. 502985557
Encompass_Preferred
A secondary bile acid produced from α-muricholic acid and β-muricholic acid; metabolite of lithocholic acid; prevents gallstones in hamsters; does not resolve gallstones in prairie dogs and induces formation of insoluble bile acid salt gallstones; rapidly absorbed in humans
Catalog No. 50-298-5557 Supplier Cayman Chemical Supplier No. 202901 MG
A secondary bile acid produced from α-muricholic acid and β-muricholic acid; metabolite of lithocholic acid; prevents gallstones in hamsters; does not resolve gallstones in prairie dogs and induces formation of insoluble bile acid salt gallstones; rapidly absorbed in humans