Learn More
ATCC Murine hepatitis virus; Strain: MHV-A59
Supplier:  ATCC VR764
Encompass
Murine hepatitis virus strain MHV-A59 is propagated in NCTC clone 1469 [derivative of NCTC 721] cells (ATCC CCL-9.1). This strain was isolated from a pool of liver and spleen suspension from BALB/c mice exhibiting rough coats and hunched positions. The strain has applications in respiratory disease research and infectious disease research.
Catalog No. 50-238-4587