Learn More
ATCC Murine hepatitis virus; Strain: MHV-JHM
Catalog No. 502382193
Encompass
Murine hepatitis virus strain MHV-JHM is propagated in NCTC clone 1469 [derivative of NCTC 721] cells (ATCC CCL-9.1). This strain was isolated from animal tissue.
Catalog No. 50-238-2193 Supplier ATCC Supplier No. VR765
Murine hepatitis virus strain MHV-JHM is propagated in NCTC clone 1469 [derivative of NCTC 721] cells (ATCC CCL-9.1). This strain was isolated from animal tissue.