Learn More
Bioworld MURINE INTERLEUKIN-12 (RECO
Supplier:  Bioworld 220605911
Encompass_Preferred
MURINE INTERLEUKIN-12 (RECOMBINANT): Recombinant Murine Interleukin-12 Source: Insect cells Background: Interleukin 12 (IL-12) is a cytokine with important regulatory functions bridging innate and adaptive immunity. It has been propo...
Catalog No. 50-253-691