Learn More
Invivogen J774-Dual Cells, 3-7 x 10e6 cells
SDP
Catalog No. NC1195147
Encompass
J774-Dual cells have been derived from the mouse J774.1 macrophage-like cell line by stable integration of two inducible reporter constructs.
Catalog No. NC1195147 Supplier Invivogen Supplier No. J774DNFIS
J774-Dual cells have been derived from the mouse J774.1 macrophage-like cell line by stable integration of two inducible reporter constructs.