Learn More
Medchemexpress LLC Muristerone A | 38778-30-2 | MFCD00056450 | 98.9% | 496.63 | C27H44O8 | 250 UG
SDP
Catalog No. 5000244635
Encompass_Preferred
Muristerone A is a phytoecdysteroid analog of ecdysone that acts as a potent agonist of the ecdysteroid receptor (reported Kd ≈ 1 nM). It is supplied as a high-purity research reagent for use in in vitro biochemical and cell-based assays. The compound has molecular formula C27H44O8 and molecular weight 496.63 g/mol; solubility in DMSO is approximately 1.5 mg/mL.
Catalog No. 50-002-44635 Supplier Medchemexpress LLC Supplier No. HY103273250UG
Muristerone A is a phytoecdysteroid analog of ecdysone that acts as a potent agonist of the ecdysteroid receptor (reported Kd ≈ 1 nM). It is supplied as a high-purity research reagent for use in in vitro biochemical and cell-based assays. The compound has molecular formula C27H44O8 and molecular weight 496.63 g/mol; solubility in DMSO is approximately 1.5 mg/mL.