Learn More
Abcam Muristerone A, EcR agonist, 1MG
Catalog No. 502705897
Encompass_Preferred
MW 496.6 Da, Purity >95%. Ecdysone receptor (EcR) agonist. Ecdysone (ab142425) analog. Potent ecdysone-inducible gene expression inducer. Stimulates Bcl-XL mRNA transcription. Inhibits TRAIL- and hFasL-induced apoptosis. Induces NGF release from fibroblasts.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5897 Supplier Abcam Supplier No. AB1443091MG
MW 496.6 Da, Purity >95%. Ecdysone receptor (EcR) agonist. Ecdysone (ab142425) analog. Potent ecdysone-inducible gene expression inducer. Stimulates Bcl-XL mRNA transcription. Inhibits TRAIL- and hFasL-induced apoptosis. Induces NGF release from fibroblasts.
The product is subject to the following: Abcam Restricted Use Statement