Learn More
Medchemexpress LLC Murrayafoline A 10mM 1mL
SDP
Catalog No. 5000205643
Encompass_Preferred
Murrayafoline A is a carbazole alkaloid that can be extracted from Murraya tetramera Murrayafoline A directly targets Specificity protein 1 (Sp1) thereby inhibiting NF- B and MAPK signaling pathways Murrayafoline a induces a G0/G1-phase arrest in platelet-derived growth factor (PDGF)-stimulated vascular smooth muscle cells Murrayafoline A attenuates the Wnt/ -catenin pathway by promoting the degradation of intracellular -catenin proteins Murrayafoline A enhances the contraction of rat ventricular myocytes and L-type calcium current by activating protein kinase C Murrayafoline A inhibits LPS (HY-D1056)-induced neuroinflammation in vivo Murrayafoline A can be used for the study of inflammation vascular complications and colon cancer[1][2][3][4]
Catalog No. 50-002-05643 Supplier Medchemexpress LLC Supplier No. HYW10028710MM/1ML
Murrayafoline A is a carbazole alkaloid that can be extracted from Murraya tetramera Murrayafoline A directly targets Specificity protein 1 (Sp1) thereby inhibiting NF- B and MAPK signaling pathways Murrayafoline a induces a G0/G1-phase arrest in platelet-derived growth factor (PDGF)-stimulated vascular smooth muscle cells Murrayafoline A attenuates the Wnt/ -catenin pathway by promoting the degradation of intracellular -catenin proteins Murrayafoline A enhances the contraction of rat ventricular myocytes and L-type calcium current by activating protein kinase C Murrayafoline A inhibits LPS (HY-D1056)-induced neuroinflammation in vivo Murrayafoline A can be used for the study of inflammation vascular complications and colon cancer[1][2][3][4]