Learn More
Biotang Inc Adiponectin, Mouse Anti Human, Acrp30 Antibody, Adiponectin antibody has been tested by ELISA and Western blot analysis to assure specificity and reactivity. Recommended dilution range for Western blot analysis is 1:250 ~ 1:1,000.
Catalog No. 50151103 Shop All BIOTANG Inc Products
Encompass_Preferred
Adiponectin, Mouse Anti Human, Acrp30 Antibody, Adiponectin antibody has been tested by ELISA and Western blot analysis to assure specificity and reactivity. Recommended dilution range for Western blot analysis is 1:250 ~ 1:1,000.
Catalog No. 50-151-103 Supplier Biotang Inc Supplier No. BTANT232
Adiponectin, Mouse Anti Human, Acrp30 Antibody, Adiponectin antibody has been tested by ELISA and Western blot analysis to assure specificity and reactivity. Recommended dilution range for Western blot analysis is 1:250 ~ 1:1,000.