Learn More
Biotang Inc INK128, Free Base, 5MG > 99%, M.W.309.33, C15H15N7O. potent mTOR kinase inhibitor with a Ki of 1.4 nM.
Catalog No. 50151360 Shop All BIOTANG Inc Products
Encompass_Preferred
INK128, Free Base, 5MG > 99%, M.W.309.33, C15H15N7O. potent mTOR kinase inhibitor with a Ki of 1.4 nM.
Catalog No. 50-151-360 Supplier Biotang Inc Supplier No. BTANT021
INK128, Free Base, 5MG > 99%, M.W.309.33, C15H15N7O. potent mTOR kinase inhibitor with a Ki of 1.4 nM.