Learn More
Biotang Inc Mouse E-Cadherin,E-Cad ELISA Kit, 96T
Catalog No. 50154170 Shop All BIOTANG Inc Products
Encompass_Preferred
Mouse E-Cadherin,E-Cad ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.
Catalog No. 50-154-170 Supplier Biotang Inc Supplier No. M7891
Mouse E-Cadherin,E-Cad ELISA Kit, 96T. Lead time of this kit is 1-2 weeks. Shipped by FedEx air. Storage: 2 - 8 C. Shelf Time: 6 months. Kit Experimental Time:< 2 hs. TSZELISA brand.