Learn More
Aviva Systems Biology Corp MUS81 Antibody - middle region (ARP61406_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61406_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUS81. It was validated on Western Blot. . Alternate names: FLJ21012, FLJ44872, SLX3ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-287-368