Learn More
Bioworld MUSA (Banana) Multiplication Medium, 25 L
Supplier:  Bioworld 306301553
Encompass_Preferred
MUSA (Banana) Multiplication MediumContains Micronutrients and Macronutrients as described by Murashige & Skoog (1962) and IITA Formulation as described by Vuylsteke (1989)Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 36.36 g/LpH: 3.75–4.75Partially solube in water
Catalog No. 50-254-879