Learn More
Medchemexpress LLC Muscarine iodide | 24570-49-8 | 97.0% | 301.17 g/mol | C9H20INO2 | 1 MG
SDP
Catalog No. 5000242169
Encompass_Preferred
Muscarine iodide is the iodide salt of muscarine and a prototypical muscarinic acetylcholine receptor (mAChR) agonist used as a research reagent. It is used to stimulate parasympathetic pathways and probe muscarinic receptor function in pharmacology and cell biology studies. Typical specifications include CAS 24570-49-8, formula C9H20INO2, molecular weight 301.17 g/mol, and listed purity of 97%.
  • Acts as a muscarinic acetylcholine receptor agonist
  • Used to stimulate parasympathetic responses in vitro
  • Supplied as iodide salt for consistent chemical identity
  • High listed purity supports reproducible experimental results
  • Available in small quantities suitable for analytical and cell-based assays
Catalog No. 50-002-42169 Supplier Medchemexpress LLC Supplier No. HY1076541MG
Muscarine iodide is the iodide salt of muscarine and a prototypical muscarinic acetylcholine receptor (mAChR) agonist used as a research reagent. It is used to stimulate parasympathetic pathways and probe muscarinic receptor function in pharmacology and cell biology studies. Typical specifications include CAS 24570-49-8, formula C9H20INO2, molecular weight 301.17 g/mol, and listed purity of 97%.
  • Acts as a muscarinic acetylcholine receptor agonist
  • Used to stimulate parasympathetic responses in vitro
  • Supplied as iodide salt for consistent chemical identity
  • High listed purity supports reproducible experimental results
  • Available in small quantities suitable for analytical and cell-based assays