Learn More
Medchemexpress LLC Muscarine iodide | 24570-49-8 | MFCD18427991 | 97.0% | 301.17 g·mol⁻1 | C9H20INO2 | 5 MG
SDP
Catalog No. 5000240115
Encompass_Preferred
Muscarine iodide is a muscarinic acetylcholine receptor (mAChR) agonist used as a pharmacological tool to study muscarinic receptor function, signaling, and parasympathetic system responses. The compound is supplied as a characterized solid with reported purity and molecular properties to support reproducible experimental work.
  • Prototype muscarinic receptor agonist for pharmacological studies.
  • Reported purity 97.0% for consistent experimental performance.
  • Molecular weight 301.17 g·mol⁻1 and formula C9H20INO2 documented.
  • Available in small pack sizes suitable for assay development.
  • Provided with product documentation to support research use.
Catalog No. 50-002-40115 Supplier Medchemexpress LLC Supplier No. HY1076545MG
Muscarine iodide is a muscarinic acetylcholine receptor (mAChR) agonist used as a pharmacological tool to study muscarinic receptor function, signaling, and parasympathetic system responses. The compound is supplied as a characterized solid with reported purity and molecular properties to support reproducible experimental work.
  • Prototype muscarinic receptor agonist for pharmacological studies.
  • Reported purity 97.0% for consistent experimental performance.
  • Molecular weight 301.17 g·mol⁻1 and formula C9H20INO2 documented.
  • Available in small pack sizes suitable for assay development.
  • Provided with product documentation to support research use.