Learn More
Abcam Muscimol, GABAA agonist, 5MG
Catalog No. NC0796509
Encompass_Preferred
MW 114.1 Da, Purity >99%. Potent, selective GABAA agonist. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0796509 Supplier Abcam Supplier No. AB1200945MG
MW 114.1 Da, Purity >99%. Potent, selective GABAA agonist. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement