Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscimol, >98%, 93.4%, For TLC analysis, MP Biomedicals™

Catalog No. ICN19533605 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19533605 5 mg
ICN19533610 10 mg
ICN19533625 25 mg
ICN19533601 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19533605 Supplier MP Biomedicals, Inc Supplier No. 0219533605
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2763-96-4
Molecular Formula C4H6N2O2
Molecular Weight (g/mol) 114.10
MDL Number MFCD00057894
InChI Key ZJQHPWUVQPJPQT-UHFFFAOYSA-N
Synonym muscimol, pantherine, agarin, agarine, 3 2h-isoxazolone, 5-aminomethyl, 5-aminomethyl-3-isoxyzole, pantherin, 5-aminomethyl-3-isoxazolol, 3-hydroxy-5-aminomethylisoxazole, 5-aminomethyl-3-hydroxyisoxazole
PubChem CID 4266
ChEBI CHEBI:7035
IUPAC Name 5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one
SMILES NCC1=CC(=O)NO1

Specifications

Color White
Assay Percent Range 0.934
Quantity 5 mg
Formula Weight 114.1
Percent Purity >98%
Physical Form Powder
Chemical Name or Material Muscimol, Neurobiology
EINECSNumber 220-430-4
Recommended Storage Store at 0°C.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.