Learn More
Abcam Muscimol, GABAA agonist, 50MG
Catalog No. 502584699
Encompass_Preferred
MW 114.1 Da, Purity >99%. Potent, selective GABAA agonist. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4699 Supplier Abcam Supplier No. AB12009450MG
Add to Cart
Add to Cart
MW 114.1 Da, Purity >99%. Potent, selective GABAA agonist. Soluble in 1 ml water to give specified mM/ml concentration.
The product is subject to the following: Abcam Restricted Use Statement