Learn More
Enzo Life Sciences Muscle actin monoclonal antibody (HHF35) (Ready-to-Use) (6ml)
SDP
Supplier:  Enzo Life Sciences ENZ30931
Encompass_Preferred
Clone: HHF35. Isotype: IgG1. Immunogen: Human myocardium (proteins extracted in 2% SDS). Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Human Rabbit, Monkey. Specificity: Recognizes actin isotypes α and γ of skeletal, cardiac and smooth muscle. This antibody labels rhabdomyosarcomas, hemangiopericytomas, leiomyosarcomas and leiomyomas but does not react with non-myogenic sarcomas. It is nonreactive with other neoplasms such as carcinoma or lymphoma and with other mesenchymal and all epithelial cells, with the exception of myoepithelium. Applications: IHC (PS), WB. Purity: Affinity purified. Formulation: Liquid. Ready-to-use solution in PBS containing 0.1% sodium azide and 1% BSA.
Catalog No. 50-201-2484