Learn More
Meso Scale Discovery Muscle Injury PNL 2 rat Kit
SDP
Catalog No. 5000078843
Encompass
The Meso Scale Discovery Muscle Injury Panel 2 Rat Kit is designed to measure biomarkers associated with muscle injury and inflammation in rat serum plasma and tissue culture media The kit contains a 96-well plate coated with capture antibodies for Creatine Kinase MM Fatty Acid Binding Protein 3 and Myoglobin The kit includes detection antibodies buffers calibrators and controls needed to run the assay Results are obtained in as little as 3 5 hours using an electrochemiluminescent detection method on the MSD platform This kit allows for the sensitive multiplexed measurement of biomarkers related to muscle injury in preclinical rat studies
Catalog No. 50-000-78843 Supplier Meso Scale Discovery Supplier No. K15180C4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Muscle Injury Panel 2 Rat Kit is designed to measure biomarkers associated with muscle injury and inflammation in rat serum plasma and tissue culture media The kit contains a 96-well plate coated with capture antibodies for Creatine Kinase MM Fatty Acid Binding Protein 3 and Myoglobin The kit includes detection antibodies buffers calibrators and controls needed to run the assay Results are obtained in as little as 3 5 hours using an electrochemiluminescent detection method on the MSD platform This kit allows for the sensitive multiplexed measurement of biomarkers related to muscle injury in preclinical rat studies