Learn More
Meso Scale Discovery Muscle Injury PNL 2 rat Kit
SDP
Catalog No. 5000082448
Encompass
The Meso Scale Discovery Muscle Injury Panel 2 Rat Kit is designed to measure biomarkers associated with muscle injury and inflammation in rat serum plasma and tissue culture media The multiplex immunoassay kit contains a 96-well plate coated with capture antibodies for Creatine Kinase MM Fatty Acid Binding Protein 3 and Myoglobin The kit includes detection antibodies calibrators controls 10X Diluent Read Buffer and Plate Seals This product allows for the simultaneous quantitative measurement of multiple muscle injury biomarkers from a single sample The electrochemiluminescent detection method provides high sensitivity wide dynamic range and low sample consumption
Catalog No. 50-000-82448 Supplier Meso Scale Discovery Supplier No. K15180C1
The Meso Scale Discovery Muscle Injury Panel 2 Rat Kit is designed to measure biomarkers associated with muscle injury and inflammation in rat serum plasma and tissue culture media The multiplex immunoassay kit contains a 96-well plate coated with capture antibodies for Creatine Kinase MM Fatty Acid Binding Protein 3 and Myoglobin The kit includes detection antibodies calibrators controls 10X Diluent Read Buffer and Plate Seals This product allows for the simultaneous quantitative measurement of multiple muscle injury biomarkers from a single sample The electrochemiluminescent detection method provides high sensitivity wide dynamic range and low sample consumption