Learn More
Meso Scale Discovery Muscle Injury PNL 3 mouse Kit
SDP
Catalog No. 5000082449
Encompass
The Meso Scale Discovery Muscle Injury Panel 3 Mouse Kit is designed to measure biomarkers associated with muscle injury and inflammation in mouse samples The kit contains a 96-well plate coated with capture antibodies for mouse Myoglobin Fatty Acid Binding Protein 3 and Creatine Kinase MM It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes all necessary reagents and can process up to 40 samples in duplicate It is intended for research use only
Catalog No. 50-000-82449 Supplier Meso Scale Discovery Supplier No. K15186C4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery Muscle Injury Panel 3 Mouse Kit is designed to measure biomarkers associated with muscle injury and inflammation in mouse samples The kit contains a 96-well plate coated with capture antibodies for mouse Myoglobin Fatty Acid Binding Protein 3 and Creatine Kinase MM It utilizes electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes all necessary reagents and can process up to 40 samples in duplicate It is intended for research use only