Learn More
Biocare Medical, LLC Muscle Specific Actin Antibody - Concentrate, 0.1 ml
SDP
Supplier:  Biocare Medical, LLC CM079A
Encompass_Preferred
The Muscle Specific Actin antibody [HHF35] recognizes muscle specific alpha and gamma actin isomers. It does not react with non-muscle actin. Studies have shown that it recognizes the alpha actin from cardiac, skeletal and smooth muscle sources. It does not react with beta or non-smooth muscle gamma actin isomers. The antibody labels leiomyoma, leiomyosarcoma, angiomyolipoma and rhabdomyosarcoma. It does not label melanoma or lymphoma. A study has suggested [HHF35] aids the differential diagnosis of Collagenous Spherulosis and Adenoid-Cystic Carcinoma of the breast.
Catalog No. 50-824-92