Learn More
Enzo Life Sciences Muscone (500mg). CAS: 541-91-3
SDP
Supplier:  Enzo Life Sciences BMLEI1610500
Encompass_Preferred
Induces rat liver microsomal cytochrome P-450. Alternative name: 3-Methylcyclopentadecanone. Purity: ≥99% (TLC). Solubility: Soluble in 100% ethanol (25mg/ml). Insoluble in water. Long Term Storage: Ambient.
Catalog No. 50-201-0580