Learn More
Medchemexpress LLC Muscone | 541-91-3 | 238.41 | 1 ML
SDP
Catalog No. 5000388366
Encompass_Preferred
Muscone is identified as the primary active monomer derived from traditional Chinese medicine musk. It functions by inhibiting NF-κB and NLRP3 inflammasome activation, leading to a significant reduction in inflammatory cytokines such as IL-1β, TNF-α, and IL-6. This action ultimately contributes to improved cardiac function and increased survival rates.
  • Inhibits NF-κB activation
  • Inhibits NLRP3 inflammasome activation
  • Decreases inflammatory cytokines (IL-1β, TNF-α, and IL-6)
  • Improves cardiac function
  • Increases survival rate
Catalog No. 50-003-88366 Supplier Medchemexpress LLC Supplier No. HYN063310MM/1ML
Add to Cart
Add to Cart
Muscone is identified as the primary active monomer derived from traditional Chinese medicine musk. It functions by inhibiting NF-κB and NLRP3 inflammasome activation, leading to a significant reduction in inflammatory cytokines such as IL-1β, TNF-α, and IL-6. This action ultimately contributes to improved cardiac function and increased survival rates.
  • Inhibits NF-κB activation
  • Inhibits NLRP3 inflammasome activation
  • Decreases inflammatory cytokines (IL-1β, TNF-α, and IL-6)
  • Improves cardiac function
  • Increases survival rate