Learn More
Medchemexpress LLC Muscone | 541-91-3 | 238.41 | 25 MG
SDP
Catalog No. 5000389558
Encompass_Preferred
Muscone is the primary active monomer found in traditional Chinese medicine musk. It functions by inhibiting NF-κB and NLRP3 inflammasome activation. Muscone has been shown to significantly reduce the levels of inflammatory cytokines (IL-1β, TNF-α, and IL-6), leading to improved cardiac function and increased survival rates.
  • Main active monomer of traditional Chinese medicine musk.
  • Inhibits NF-κB and NLRP3 inflammasome activation.
  • Remarkably decreases levels of inflammatory cytokines (IL-1β, TNF-α, and IL-6).
  • Improves cardiac function and survival rate.
  • Downregulates LPS-induced inflammatory cytokines in bone marrow-derived macrophages (BMDMs) in vitro.
  • Considered a promising drug for post-myocardial infarction (MI) treatment.
Catalog No. 50-003-89558 Supplier Medchemexpress LLC Supplier No. 25MGHYN0633
Add to Cart
Add to Cart
Muscone is the primary active monomer found in traditional Chinese medicine musk. It functions by inhibiting NF-κB and NLRP3 inflammasome activation. Muscone has been shown to significantly reduce the levels of inflammatory cytokines (IL-1β, TNF-α, and IL-6), leading to improved cardiac function and increased survival rates.
  • Main active monomer of traditional Chinese medicine musk.
  • Inhibits NF-κB and NLRP3 inflammasome activation.
  • Remarkably decreases levels of inflammatory cytokines (IL-1β, TNF-α, and IL-6).
  • Improves cardiac function and survival rate.
  • Downregulates LPS-induced inflammatory cytokines in bone marrow-derived macrophages (BMDMs) in vitro.
  • Considered a promising drug for post-myocardial infarction (MI) treatment.