Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscone, ≥95%, MP Biomedicals™

Catalog No. ICN15918680 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15918680 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15918680 Supplier MP Biomedicals, Inc Supplier No. 0215918680

missing translation for 'validMainframeMsg'

Muscone, originally collected from male musk deer ventral glands, is a potent antiinflammatory agent that has been reported to reduce proinflammatory cytokines in vitro

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-91-3
Molecular Formula C16H30O
Molecular Weight (g/mol) 238.42
MDL Number MFCD00211114
InChI Key ALHUZKCOMYUFRB-UHFFFAOYNA-N
Synonym muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanone
PubChem CID 10947
IUPAC Name 3-methylcyclopentadecan-1-one
SMILES CC1CCCCCCCCCCCCC(=O)C1

Specifications

Color Colorless
Density 0.84g/mL
Assay Percent Range ≥95%
Quantity 100 mg
Formula Weight 238.43
Physical Form Liquid
Chemical Name or Material Muscone
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.