Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Muscone 97.0+%, TCI America™
SDP

Catalog No. M2611200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2611200MG 200 mg
M26111G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2611200MG Supplier TCI America Supplier No. M2611200MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 541-91-3
Molecular Formula C16H30O
Molecular Weight (g/mol) 238.42
MDL Number MFCD00211114
InChI Key ALHUZKCOMYUFRB-UHFFFAOYNA-N
Synonym muscone, 3-methylcyclopentadecanone, muskone, methylexaltone, moschus ketone, cyclopentadecanone, 3-methyl, 3-methyl-1-cyclopentadecanone, 5-methyl-1-cyclopentadecanone, fema no. 3434, 3-methyl-cyclopentadecanone
PubChem CID 10947
IUPAC Name 3-methylcyclopentadecan-1-one
SMILES CC1CCCCCCCCCCCCC(=O)C1

Specifications

Melting Point -15°C
Color Colorless
Boiling Point 95°C
Quantity 200 mg
Formula Weight 238.42
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Muscone
EINECSNumber (3)-4630
RTECSNumber GY0950000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.