Learn More
Electron Microscopy Sciences V-2 Quality, 15 x 15 mm, 0.15- 0.21 mm thick
SDP
Catalog No. 50949069
Encompass
EMS Mica Sheets offer a clean surface for EM applications, carbon filming & particle spraying, as well as for AFM applications. There are 2 types of mica: muscovite & phlogopite. Generally, one differs from the other by color (Muscovite is Ruby, Green or White; Phlogopite is Amber, Yellow, or Silver). The max operating temp for Muscovite is about 500-600 deg C & for Phlogopite is about 800-900 deg C. Our line consists of High Quality Muscovite Mica. This mica peels off very thin up to 0.0001" uniform layers, exposing "virgin" mica upon splitting.
Muscovite, potash type mica, sometimes known as granitic mica, is the best of all micas in dielectric strength, perfection of cleavage, & transparency. It has the chem formula: H2KAI3(Si04)3. Clear & Slightly Stained. Hard, uniform color, nearly flat & slight crystallographic discoloration. Very slight air inclusions & not more than 1/4 of usable area.
Quality: V-2
Size: 15 x 15 mm
Thickness: 0.15- 0.21 mm
Catalog No. 50-949-069 Supplier Electron Microscopy Sciences Supplier No. 7185715
EMS Mica Sheets offer a clean surface for EM applications, carbon filming & particle spraying, as well as for AFM applications. There are 2 types of mica: muscovite & phlogopite. Generally, one differs from the other by color (Muscovite is Ruby, Green or White; Phlogopite is Amber, Yellow, or Silver). The max operating temp for Muscovite is about 500-600 deg C & for Phlogopite is about 800-900 deg C. Our line consists of High Quality Muscovite Mica. This mica peels off very thin up to 0.0001" uniform layers, exposing "virgin" mica upon splitting.
Muscovite, potash type mica, sometimes known as granitic mica, is the best of all micas in dielectric strength, perfection of cleavage, & transparency. It has the chem formula: H2KAI3(Si04)3. Clear & Slightly Stained. Hard, uniform color, nearly flat & slight crystallographic discoloration. Very slight air inclusions & not more than 1/4 of usable area.
Quality: V-2
Size: 15 x 15 mm
Thickness: 0.15- 0.21 mm