Learn More
Electron Microscopy Sciences V5 Quality, 15 x 15 mm, 0.26- 0.31 mm thick
SDP
Catalog No. 50949248
Encompass
EMS Mica Sheets offer a clean surface for EM applications, carbon filming & particle spraying, as well as for AFM applications. There are 2 types of mica: muscovite & phlogopite. Generally, one differs from the other by color (Muscovite is Ruby, Green or White; Phlogopite is Amber, Yellow, or Silver). The maxoperating temp.for Muscovite is about 500-600 degree C & for Phlogopite is about 800-900 degree C. Our line consists of High Quality Muscovite Mica. This mica peels off very thin up to 0.0001" uniform layers, exposing "virgin" mica upon splitting.
Muscovite, potash type mica, sometimes known as granitic mica, is the best of all micas in dielectric strength, perfection of cleavage, & transparency. It has the chemical formula: H2KAI3(Si04)3. Stained A Quality. Hard, med. air inclusions uniformly distributed in usable area; slight green vegetable stains, med. waviness, & heavy waves if specified.
Quality: V5
Size: 15 x 15 mm
Thickness: 0.26- 0.31 mm
Catalog No. 50-949-248 Supplier Electron Microscopy Sciences Supplier No. 7185015
EMS Mica Sheets offer a clean surface for EM applications, carbon filming & particle spraying, as well as for AFM applications. There are 2 types of mica: muscovite & phlogopite. Generally, one differs from the other by color (Muscovite is Ruby, Green or White; Phlogopite is Amber, Yellow, or Silver). The maxoperating temp.for Muscovite is about 500-600 degree C & for Phlogopite is about 800-900 degree C. Our line consists of High Quality Muscovite Mica. This mica peels off very thin up to 0.0001" uniform layers, exposing "virgin" mica upon splitting.
Muscovite, potash type mica, sometimes known as granitic mica, is the best of all micas in dielectric strength, perfection of cleavage, & transparency. It has the chemical formula: H2KAI3(Si04)3. Stained A Quality. Hard, med. air inclusions uniformly distributed in usable area; slight green vegetable stains, med. waviness, & heavy waves if specified.
Quality: V5
Size: 15 x 15 mm
Thickness: 0.26- 0.31 mm